Influenza A H7N9 (A/Anhui/1/2013) Recombinant Hemagglutinin/HA (HA1+HA2, cleavage) Protein (His Tag) (from HEK293 Cells)
Catalog # 103632-844
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H7N9
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:EPI439507
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of Influenza A virus (A/Anhui/1/2013(H7N9)) comprises 517 amino acids and has a predicted molecular mass of 57.6 kDa. The apparent molecular mass of the protein is approximately 43 and 27 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Val524
- Shipping Temperature:Ambient
- Cat. No.:103632-844
Specifications
About this item
A DNA sequence encoding the Influenza A virus (A/Anhui/1/2013(H7N9)) hemagglutinin (EPI439507) (Met1-Val524) (cleavage between Arg339 and Gly440, HA1+HA2) was expressed with a C-terminal polyhistidine tag.