Human Recombinant IL-24/MDA-7 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103887-588
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Gene ID:NP_001172085.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:C49A|ST16|MDA7|IL10B|MOB5|FISP
- Protein/Peptide Name:IL24
- Purity:>80% as determined by SDS-PAGE
- Molecular Weight:The recombinant human IL24 consists 389 amino acids and predicts a molecular mass of 44.6 kDa.
- Sequence:Met1-Leu207
- Shipping Temperature:Ambient
- Cat. No.:103887-588
Specifications
About this item
A DNA sequence encoding the human IL24 (NP_001172085.1) (Met1-Leu207) was expressed with the Fc region of mouse IgG1 at the C-terminus.