Specifications
- Description:Corning® X-MINI® pressor for Corning X-MINI selection kits
- Environmentally Preferable:
- Cat. No.:76470-988
- Packaging:1/pack, 1 pack/case
Specifications
About this item
The Corning X-MINI Pressor is designed to be used exclusively with the Corning X-MINI Selection Kits.
The Corning X-MINI Pressor allows for the degassing of microbubble-targeted cells. It accommodates a capped 10 ml syringe.
The pressure generated using the X-MINI Pressor is sufficient to isolate up to 1×10⁷ cells, and one syringe can process up to 3 ml of cell suspension at a time.