Order Entry
United States
ContactUsLinkComponent
Cynomolgus Recombinant CD73/NT5E Protein (His Tag) (from HEK293 Cells)
Cynomolgus Recombinant CD73/NT5E Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
Cynomolgus Recombinant CD73/NT5E Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to hydrolyze the 5’-phosphate group from the substrate adenosine-5’-monophosphate (AMP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996).
    The specific activity is >20,000 pmol/min/µg.
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    eN|E5NT|NTE|NT5|CALJA|eNT|NT|CD73
  • Protein/Peptide Name:
    NT5E
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant cynomolgus NT5E comprises 532 amino acids and has a calculated molecular mass of 59.2 KDa. The apparent molecular mass of it is approximately 59 KDa respectively in SDS-PAGE.
  • Sequence:
    Met1-Lys547
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together