Rat Recombinant CD70/CD27L/TNFSF7 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-396
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized mouse CD27-His (Cat:50110-M08H) at 10 μg/ml (100 μl/well) can bind rat Fc-CD70 with a linear range of 0.16-1.25 μg/ml.
- Gene ID:D3ZGU3
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD27LG|TNFSF7|CD27L
- Protein/Peptide Name:CD70
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat CD70 comprises 410 amino acids and predicts a molecular mass of 45 kDa. The apparent molecular mass of the recombinant protein is approximately 55 kDa in SDS-PAGE under reducing conditions due to glycosylation.
- Sequence:Gln46-Pro195
- Shipping Temperature:Ambient
- Cat. No.:103666-396
Specifications
About this item
A DNA sequence encoding the rat CD70 (D3ZGU3) (Gln46-Pro195) was expressed, fused with Fc region of human IgG1 at the N-terminus.