Order Entry
United States
ContactUsLinkComponent
Rat Recombinant Ephrin-B3/EFNB3 Protein (His Tag) (from HEK293 Cells)
Rat Recombinant Ephrin-B3/EFNB3 Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-346
Supplier:  Sino Biological
CAS Number:  
undefined
Rat Recombinant Ephrin-B3/EFNB3 Protein (His Tag) (from HEK293 Cells)
Catalog # 103666-346
Supplier:  Sino Biological
Supplier Number:  80112-R08H-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized rat EFNB3-His at 10 μg/ml (100 μl/well) can bind biotinylated mouse EphB3-His (Cat:50581-M08H), The EC50 of biotinylated mouse EphB3-His (Cat:50581-M08H) is 17.4-40.6 ng/ml.
  • Gene ID:
    G3V7D4
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Epl8|LERK-8|NLERK-2|EFL-6|ELF-3|Elk-L3
  • Protein/Peptide Name:
    EFNB3
  • Purity:
    >85% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant rat EFNB3 comprises 208 amino acids and predicts a molecular mass of 23.1 kDa. The apparent molecular mass of the recombinant protein is approximately 32 kDa in SDS-PAGE under reducing conditions due to glycosylation.
  • Sequence:
    Met1-Ser224
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103666-346

Specifications

About this item

A DNA sequence encoding the rat EFNB3 (G3V7D4) (Met1-Ser224) was expressed, fused with a polyhistidine tag at the C-terminus.