Rat Recombinant GPT2 Protein (His Tag) (from Baculovirus Insect Cells)
Catalog # 103666-204
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Rat
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_001012057.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Gpt2
- Protein/Peptide Name:Gpt2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rat Gpt2 comprises 532 amino acids with a predicted molecular mass of 55 kDa. It migrates as an approximately 48 kDa band in reduced SDS-PAGE.
- Sequence:Met1-Ser522
- Shipping Temperature:Ambient
- Cat. No.:103666-204
Specifications
About this item
A DNA sequence encoding the rat Gpt2 (NP_001012057.1) (Met 1-Ser 522) with a C-terminal polyhistidine tag was expressed.