Order Entry
United States
ContactUsLinkComponent
Canine Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Canine Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-176
Supplier:  Sino Biological
CAS Number:  
undefined
Canine Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103666-176
Supplier:  Sino Biological
Supplier Number:  70109-D02H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Dog
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    Immobilized PD-1 Protein, Canine, Recombinant (hFc Tag)(Cat:70109-D02H) at 2 μg/ml (100 μl/well) can bind PD-L1 Protein, Canine, Recombinant (hFc Tag), Biotinylated(Cat:70110-D02H-B),the EC50 is 300-1200 ng/ml.
  • Gene ID:
    XP_543338.3
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    PD-1|hPD-1|hPD-l|CD279|hSLE1|PD1|SLEB2
  • Protein/Peptide Name:
    PDCD1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant canine PDCD1 consists 383 amino acids and predicts a molecular mass of 43 kDa.
  • Sequence:
    Met1-Leu169
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103666-176

Specifications

About this item

A DNA sequence encoding the canine PDCD1 (XP_543338.3) (Met1-Leu169) was expressed with the Fc region of human IgG1 at the C-terminus.