Dengue virus DENV-2 (strain New Guinea C) Recombinant NS5 (methyltransferase domain)/Nonstructural 5 Protein (His Tag) (from E. coli)
Catalog # 103633-972
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:DENV
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:AAC59275.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:DENV-NS5A
- Protein/Peptide Name:DENV-NS5A
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Dengue virus (type 2, strain New Guinea C) NS5 consists 314 amino acids and predicts a molecular mass of 35.5 kDa.
- Sequence:Gly2492-Gln2787
- Shipping Temperature:Ambient
- Cat. No.:103633-972
Specifications
About this item
A DNA sequence encoding the Dengue virus (type 2, strain New Guinea C) NS5 (AAC59275.1) (Gly2492-Gln2787) was expressed with a polyhistidine tag at the N-terminus.