Influenza A H5N2 (A/ostrich/South Africa/AI1091/2006) Recombinant Hemagglutinin (HA1 Subunit) Protein (His Tag) (from HEK293 Cells)
103633-906
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Influenza A
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ABQ24010.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The secreted recombinant HA1 subunit of influenza A H5N1 HA (A/ostrich/SouthAfrica/AI1091/2006 (H5N2)) comprises 338 amino acids and has a predicted molecular mass of 38.2 kDa. As a result of glycosylation, it migrates as an approximately 50 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Arg355
- Shipping Temperature:Ambient
- Cat. No.:103633-906
A DNA sequence encoding the N-terminal segment (Met 1-Arg 355) of the influenza hemagglutinin (A/ostrich/South Africa/AI1091/2006 (H5N2)) (ABQ24010.1), termed as HA1, was fused with a polyhistidine tag at the C-terminus