Human Recombinant TMIGD2 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103887-282
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized human HHLA2h (Cat:16139-H08H) at 10 μg/ml (100 μl/well) can bind human TMIGD2-Fc3 (Cat:13820-H02H), the EC50 of human TMIGD2-Fc3 (Cat:13820-H02H) is 50-200 ng/ml.
- Gene ID:NP_653216.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TPC1|TMIGD2|IGPR1|IGPR-1
- Protein/Peptide Name:TMIGD2
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human TMIGD2 consists of 368 amino acids and predicts a molecular mass of 40.9 kDa.
- Sequence:Met1-Gly150
- Shipping Temperature:Ambient
- Cat. No.:103887-282
Specifications
About this item
A DNA sequence encoding the human TMIGD2 ( NP_653216.2) (Met1-Gly150) was expressed with the Fc region of human IgG1 at the C-terminus.