Human Recombinant Nectin 3 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103886-324
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized PVRL1/NECTIN1 Protein, Human, Recombinant (His Tag)(Cat:11611-H08H) at 2 μg/ml (100 μl/well) can bind Nectin 3 Protein, Human, Recombinant (ECD, hFc Tag)(Cat:10852-H02H), the EC50 of Nectin 3 Protein, Human, Recombinant (ECD, hFc Tag)is 30-140 ng/ml.
- Gene ID:NP_056295.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:PRR3|nectin-3|CD113|CDw113|PPR3|PVRR3|PVRL3|DKFZp566B0846|FLJ90624
- Protein/Peptide Name:NECTIN3
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human NECTIN3 consists of 581 amino acids and predicts a molecular mass of 64.4 kDa.
- Sequence:Met1-Asp400
- Shipping Temperature:Ambient
- Cat. No.:103886-324
Specifications
About this item
A DNA sequence encoding the human NECTIN3 (NP_056295.1) (Met1-Asp400) was expressed with the Fc region of human IgG1 at the C-terminus.