Order Entry
United States
ContactUsLinkComponent
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Catalog # 76483-876
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 532
Catalog # 76483-876
Supplier:  AAT Bioquest
Supplier Number:  23117
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Alexa Fluor® 532
  • Protein/Peptide Type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Size:
    300 tests
  • Protein Synonyms:
    Amanita phalloides
  • Protein/Peptide Name:
    Phalloidin
  • Cat. No.:
    76483-876

Specifications

About this item

This orange fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 532-labeled phalloidin) selectively binds to F-actins.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.