Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 488
Catalog # 76483-872
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Vial
- Conjugation:Alexa Fluor® 488
- Protein/Peptide Type:Native
- Source:Eukaryotic cells
- Species:Amanita phalloides
- Size:300 tests
- Environmentally Preferable:
- Protein Synonyms:Amanita phalloides
- Protein/Peptide Name:Phalloidin
- Cat. No.:76483-872
Specifications
About this item
This green fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 488-labeled phalloidin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Caution: For research use only.