SevenDirect™ SD23 pH/Conductivity Meter
About this item
The pH/conductivity meter SevenDirect™ SD23 provides accurate pH and conductivity measurements for almost any sample across a wide range of applications. The EasyPlace electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
- Intuitive user interface using 7" touchscreen
- On-screen instructions guide through the calibration process
- EasyPlace™ electrode arm ensures precise placement of the sensor
- Feature a closed connector compartment to prevent corrosion
- A replacable protective cover keeps the instrument safe from dust and spills (IP 54 in-use)
- Instruments support GLP compliance by storing key measurement data
The intuitive user interface of the generous 7" touchscreen facilitates the measurement process by avoiding unnecessary steps and focusing on the most important information. The SevenDirect™ has a guided verification process that provides a straightforward way to check system performance with a clear pass/fail indication. The meter has a replaceable protective cover to keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use).
Delivery information: The meter-only model includes EasyPlace™ electrode holder, protective cover, Get-Started guide and power supply. The kits also include the InLab expert Pro-ISM, conductivity sensor InLab 731-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2×2 conductivity standards (1413 μs/cm and 12.88 ms/cm), in addition to the meter-only standard accessories.
The pure H₂O kit includes the InLab Pure Pro-ISM, conductivity sensor InLab 741-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2 conductivity standards (84 μs/cm).
3 Options Available Below
Product Details & Documents
The pH/conductivity meter SevenDirect™ SD23 provides accurate pH and conductivity measurements for almost any sample across a wide range of applications. The EasyPlace electrode arm and protective cover are included with the instrument. The benchtop meter's ease of use, data handling options and robustness make it ideal for all industries.
- Intuitive user interface using 7" touchscreen
- On-screen instructions guide through the calibration process
- EasyPlace™ electrode arm ensures precise placement of the sensor
- Feature a closed connector compartment to prevent corrosion
- A replacable protective cover keeps the instrument safe from dust and spills (IP 54 in-use)
- Instruments support GLP compliance by storing key measurement data
The intuitive user interface of the generous 7" touchscreen facilitates the measurement process by avoiding unnecessary steps and focusing on the most important information. The SevenDirect™ has a guided verification process that provides a straightforward way to check system performance with a clear pass/fail indication. The meter has a replaceable protective cover to keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use).
Delivery information: The meter-only model includes EasyPlace™ electrode holder, protective cover, Get-Started guide and power supply. The kits also include the InLab expert Pro-ISM, conductivity sensor InLab 731-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2×2 conductivity standards (1413 μs/cm and 12.88 ms/cm), in addition to the meter-only standard accessories.
The pure H₂O kit includes the InLab Pure Pro-ISM, conductivity sensor InLab 741-ISM, 3×2 pH buffer (pH 4.01, 7.00 and 9.21 or 10.01) sachets and 2 conductivity standards (84 μs/cm).
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 76533-434
- 76533-468
- 76533-470
- pH Resolution:
- 0.1; 0.01; 0.001
- 0.1; 0.01; 0.001
- 0.1; 0.01; 0.001
- pH Accuracy:
- 0.002
- 0.002
- 0.002
- mV resolution:
- 1; 0.1
- 1; 0.1
- 1; 0.1
- Conductivity Range:
- 0.001 μs/cm - 2000 ms/cm
- 0.001 μs/cm - 2000 ms/cm
- 0.001 μs/cm - 2000 ms/cm
- Temperature Range:
- –30...130 °C
- –30...130 °C
- –30...130 °C
Specifications
Product Family Options
Meter only
Product Information
- AvailabilityPrice
- 76533-434pH/Conductivity meter SevenDirect™ SD23Supplier Number: 30671553Specifications:
More
Specifications
Cat. No.76533-434pH Resolution0.1; 0.01; 0.001pH Accuracy0.002mV resolution1; 0.1Conductivity Range0.001 μs/cm - 2000 ms/cmTemperature Range–30...130 °C
Kits
Product Information
- AvailabilityPrice
- 76533-468pH/Conductivity meter SevenDirect™ SD23 kitSupplier Number: 30671567Specifications:
More
Specifications
Cat. No.76533-468pH Resolution0.1; 0.01; 0.001pH Accuracy0.002mV resolution1; 0.1Conductivity Range0.001 μs/cm - 2000 ms/cmTemperature Range–30...130 °C - 76533-470pH/Conductivity meter SevenDirect™ SD23 Pure H₂O kitSupplier Number: 30671568Specifications:
More
Specifications
Cat. No.76533-470pH Resolution0.1; 0.01; 0.001pH Accuracy0.002mV resolution1; 0.1Conductivity Range0.001 μs/cm - 2000 ms/cmTemperature Range–30...130 °C
Accessories
Product Information
- Catalog #DetailsAvailabilityPrice
- Parking beaker EasyPlace™Parking beaker EasyPlace™76533-476The parking beaker EasyPlace™ for the SevenDirect™ pH meter is robust and resistant to chemicals for most common lab applications. The beaker has convenient 20 mL graduations for quick and easy measurements.Log in for availabilityEach - $24.20
Product Information
- Catalog #DetailsAvailabilityPrice
- Magnetic stirrer mixMagnetic stirrer mix76533-474The powerful magnetic stirrer EasyMix™ is controlled by the SevenDirect™ pH meter helping stir the sample before or during the measurement. Stirring with the EasyMix™ reduces overall measurement error allows higher reproducibility of your results.Log in for availabilityEach - $399.00
Product Information
- Catalog #DetailsAvailabilityPrice
- Electrode arm EasyPlace™Electrode arm EasyPlace™76533-472The electrode arm EasyPlace™ is included with every SevenDirect™ pH meter. This accessory helps provide precise positioning of electrodes, a tidy workplace and streamlined calibration and measurement.Log in for availabilityEach - $335.00
Product Information
- Catalog #DetailsAvailabilityPrice
- Parking beaker insertParking beaker insert76533-478The parking beaker insert EasyPlace™ for the SevenDirect™ pH meter is secure, keeping the beaker in place while working with various lab samples.Log in for availabilityEach - $9.40
Product Information
- Catalog #DetailsAvailabilityPrice
- Sachet holder EasyPlace™Sachet holder EasyPlace™76533-480The sachet holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sachets in place. The EasyPlace™ sachet holder is made for the newly designed pH and conductivity style sachets to ensure calibration is quick and easy.Log in for availabilityEach - $49.50
Product Information
- Catalog #DetailsAvailabilityPrice
- Sensor holder EasyPlace™ singleSensor holder EasyPlace™ single76533-482The sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements.Log in for availabilityEach - $24.20
Product Information
- Catalog #DetailsAvailabilityPrice
- Sensor holder EasyPlace™ single extendedSensor holder EasyPlace™ single extended76533-486The sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements.Log in for availabilityEach - $29.70
Product Information
- Catalog #DetailsAvailabilityPrice
- Pole EasyPlace™ 30 cmPole EasyPlace™ 30 cm76533-488The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Log in for availabilityEach - $242.00
Product Information
- Catalog #DetailsAvailabilityPrice
- Sensor holder EasyPlace™ tripleSensor holder EasyPlace™ triple76533-484The sensor holder EasyPlace™ for the SevenDirect™ pH meter is extremely convenient for holding sensors in place to ensure accurate measurements.Log in for availabilityEach - $29.70
Product Information
- Catalog #DetailsAvailabilityPrice
- Pole EasyPlace™ 50 cmPole EasyPlace™ 50 cm76533-490The pole EasyPlace™ for the SevenDirect™ pH meter is attached to the base plate to hold electrodes securely and safely.Log in for availabilityEach - $287.00
Product Information
- Catalog #DetailsAvailabilityPrice
- Base plate EasyPlace™Base plate EasyPlace™76533-492The base plate EasyPlace™ for the SevenDirect™ meter is extremely secure, distributing weight evenly over the entire meter area. Meter parts such as the electrode arm will be attached to the base plate when you start to build your SevenDirect pH meter.Log in for availabilityEach - $145.00
Product Information
- Catalog #DetailsAvailabilityPrice
- Power supply 18WPower supply 18W76533-494The power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter.Log in for availabilityEach - $95.80
Product Information
- Catalog #DetailsAvailabilityPrice
- Plug set 18 W power supplyPlug set 18 W power supply76533-496The plug set power supply (18 W) can be used as an external power source for the SevenDirect™ pH meter. Additional adaptors included in the plug set.Log in for availabilityEach - $32.60
Product Information
- Catalog #DetailsAvailabilityPrice
- Top cover pole EasyPlace™Top cover pole EasyPlace™76533-498The top cover pole EasyPlace™ fits tightly over the EasyPlace™ stand for an added layer of protection when not operating the meter.Log in for availabilityEach - $24.20
Product Information
- Catalog #DetailsAvailabilityPrice
- Screws and feet set EasyPlace™Screws and feet set EasyPlace™76533-500The EasyPlace™ screws and feet set are included with the base plate for SevenDirect™ meter assembly.Log in for availabilityEach - $29.70
Product Information
- Catalog #DetailsAvailabilityPrice
- Protective cover SevenDirect™Protective cover SevenDirect™76533-502The clear, protective cover fits tightly over your SevenDirect™ meter for an added layer of protection against spills and scratches.Log in for availabilityEach - $56.65