Human Recombinant CD27 Protein (Fc Tag) (from HEK293 Cells)
103906-066
:
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_001233.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Tp55|S152|T14|TNFRSF7|S152.LPFS2|S152. LPFS2
- Protein/Peptide Name:CD27
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human CD27 consists of 411 amino acids and predicts a molecular mass of 46.1 kDa.
- Sequence:Met1-Ile192
- Shipping Temperature:Ambient
- Cat. No.:103906-066
A DNA sequence encoding the human CD27 (NP_001233.1) (Met1-Ile192) was expressed with the Fc region of human IgG1 at the C-terminus.