Dextran, M.W. 3,000 Cal-520™ labelled
Catalog # 76483-092
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Vial
- Size:1 mg
- Cat. No.:76483-092
Specifications
About this item
:
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935