Vikan® 28" Double-Blade Ultra-Hygiene Squeegees, Remco
Supplier: Remco Products
Certificates
About this item
The double-blade, non-porous rubber design of this squeegee is effective in removing water from all types of floors, including tile, epoxy, and cement. It is very efficient in large food production facilities, on walkways and expansive areas.
- Rubber blade, polypropylene block
- Seamless design
- Euro (hygienic) thread type
The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical and hygienic option.
Ordering information: Replacement blades must be ordered separately.
Frequently Bought Together