Order Entry
United States
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
Catalog # 76485-144
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, ReadiUse™ Peroxide-and Carbonyl-Free
Catalog # 76485-144
Supplier:  AAT BIOQUEST INC
Supplier Number:  60012
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    50 ml
  • Cat. No.:
    76485-144

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505