Specifications
- Description:Alpha-HP disinfectant cleaner
- Size:1.5 L
- Environmentally Preferable:
- Cat. No.:76514-344
Specifications
About this item
One product cleans and brightens surfaces safely and easily.
- Bactericidal and virucidal, no-food contact sanitizer
- Hydrogen peroxide active ingreadient breaks down to oxygen and water after use
- Excellent cleaning performance and environmental profile
- Meets daily cleaning and disinfection needs
- For use on hard, inanimate, non-porous surfaces such as floors, walls, glazed procelain and plastic surfaces
An all-in-one, multi-surface disinfectant cleaner based on proprietary accelerated hydrogen peroxide (AHP®) technology. One product cleans and brightens surfaces safely and easily. Use on walls, countertops and most other water washable hard surfaces.