ContactUsLinkComponent
Beadlite™ Rapid Colorimetric Amino Quantitation Kit for Nanoparticles, AAT Bioquest
  76485-164
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Beadlite™ Rapid Colorimetric Amino Quantitation Kit for Nanoparticles, AAT Bioquest
  76485-164
 :  5532
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Beadlite™ rapid colorimetric amino quantitation kit for nanoparticles
  • Cat. No.:
    76485-164
  • No. of tests:
    2
Surface functionalized micro- and nano-particles have been widely used in the field of nanotechnologies and biological sciences such as bio-separation and purification, assay development, and drug delivery.
  • High specificity
  • Proportional absorbance measurements
  • Quick protocol
 : For research use only.