Specifications
- Description:ReadiLink™ KLH conjugation kit
- Size:2 Reactions
- Cat. No.:76481-788
Specifications
About this item
Keyhole Limpet Hemocyanin (KLH) is one of the most commonly used carriers in the conjugation of peptides for antibody production.
- Excellent protein recovery
- Stable conjugation
- Easy and simple protocol