ContactUsLinkComponent
ReadiLink™ Protein Conjugation Stop Buffer (1 ml), AAT Bioquest
ReadiLink™ Protein Conjugation Stop Buffer (1 ml), AAT Bioquest
Catalog # 76481-784
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiLink™ Protein Conjugation Stop Buffer (1 ml), AAT Bioquest
Catalog # 76481-784
Supplier:  AAT BIOQUEST INC
Supplier Number:  5400
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    ReadiLink™ protein conjugation stop buffer
  • Size:
    1 ml
  • Cat. No.:
    76481-784
Specifications
About this item
ReadiLink™ Protein Conjugation Stop Buffer is used to stop the protein conjugation reactions that use succinimidyl ester, sulfonyl chloride or isothiocyanate.
  • Usable at room temperature
  • Stops all major protein conjugation reactions
  • Simple protocol
Caution: For research use only.