ContactUsLinkComponent
ReadiLink™ Rapid XFD488 Antibody Labeling Kit *XFD488 Same Structure to Alexa Fluor™ 488* (2×50 µg Labelings), AAT Bioquest
ReadiLink™ Rapid XFD488 Antibody Labeling Kit *XFD488 Same Structure to Alexa Fluor™ 488* (2×50 µg Labelings), AAT Bioquest
  76485-560
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
ReadiLink™ Rapid XFD488 Antibody Labeling Kit *XFD488 Same Structure to Alexa Fluor™ 488* (2×50 µg Labelings), AAT Bioquest
  76485-560
 :  1275
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    ReadiLink™ rapid XFD488 antibody labeling kit 'XFD488 same structure to Alexa Fluor™ 488'
  • Size:
    2×50 µg labelings
  • Cat. No.:
    76485-560
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
 : For research use only.