ContactUsLinkComponent
Buccutite™ FOL, NHS ester (2 µmoles), AAT Bioquest
Buccutite™ FOL, NHS ester (2 µmoles), AAT Bioquest
Catalog # 76484-958
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Buccutite™ FOL, NHS ester (2 µmoles), AAT Bioquest
Catalog # 76484-958
Supplier:  AAT BIOQUEST INC
Supplier Number:  5350
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Buccutite™ FOL, NHS ester
  • Cat. No.:
    76484-958
  • Features:
    2 µmoles
Specifications
About this item
Our Buccutite™crosslinking technology provides the most convenient and effective crosslinking method to link two biomolecules with a high conjugation yield.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol