ContactUsLinkComponent
Annexin V, FITC Labeled, 100 tests, AAT Bioquest
Annexin V, FITC Labeled, 100 tests, AAT Bioquest
Catalog # 76482-980
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Annexin V, FITC Labeled, 100 tests, AAT Bioquest
Catalog # 76482-980
Supplier:  AAT BIOQUEST INC
Supplier Number:  20030
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Annexin V, FITC Labeled
  • Cat. No.:
    76482-980
  • No. of tests:
    100
Specifications
About this item
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
  • High stability
  • In-house quality control
  • Multiple research applications
Caution: For research use only.