Amplite® Colorimetric Aldehyde Quantitation Kit , AAT Bioquest
Catalog # 76481-846
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Amplite™ colorimetric aldehyde quantitation kit
- Cat. No.:76481-846
- No. of tests:200
Specifications
About this item
Rapid and accurate measurement of aldehydes is important task for biological research, food industry, chemical research and environmental pollution surveillance.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.