ContactUsLinkComponent
Amplite® Fluorimetric Protein Quantitation Kit *Orange Fluorescence* , AAT Bioquest
Amplite® Fluorimetric Protein Quantitation Kit *Orange Fluorescence* , AAT Bioquest
Catalog # 76484-658
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Amplite® Fluorimetric Protein Quantitation Kit *Orange Fluorescence* , AAT Bioquest
Catalog # 76484-658
Supplier:  AAT BIOQUEST INC
Supplier Number:  11105
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amplite™ fluorimetric protein quantitation kit *orange fluorescence*
  • Cat. No.:
    76484-658
  • No. of tests:
    500
Specifications
About this item
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.