Order Entry
United States
Orders LinkContactUsLinkComponent
cBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest

cBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest

BSA cationized with ethylenediamine, 10 mg, Lyophilized, Non sterile, Bottle
Catalog # 76481-808
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier:   AAT Bioquest
Supplier #:  5600
cBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest
Catalog # 76481-808
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
cBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest
cBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest

BSA cationized with ethylenediamine, 10 mg, Lyophilized, Non sterile, Bottle

Price
$257.44
Log in for availability
Each (10mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

cBSA is the bovine serum albumin modified with ethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).

  • Lyophilized powder
  • Excellent quality control
  • Stronger immune response
  • 60000 molecular weight

Caution: For research use only

Specifications
  • Serum format:
    Lyophilized
  • Sterility:
    Non sterile
  • Description:
    BSA cationized with ethylenediamine
  • Size:
    10 mg
  • Packaging:
    Bottle
  • Cat. No.:
    76481-808
  • Purification:
    97%, via fractionation
  • Application:
    Use as a carrier protein for immunization
  • Storage Temperature:
    <–20 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.