Order Entry
United States
ContactUsLinkComponent
Blood Agar, 10% Sheep in Tryptic Soy Agar (TSA) Base, Hardy Diagnostics
Blood Agar, 10% Sheep in Tryptic Soy Agar (TSA) Base, Hardy Diagnostics
Catalog # 76523-382
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Blood Agar, 10% Sheep in Tryptic Soy Agar (TSA) Base, Hardy Diagnostics
Catalog # 76523-382
Supplier:  Hardy Diagnostics
Supplier Number:  A71
CAS Number:  

Specifications

  • Description:
    Prepared tryptic soy agar media
  • Dimensions:
    15×100 mm
  • Size:
    10/pack
  • Cat. No.:
    76523-382
  • Format:
    Plate

Specifications

About this item

(Tryptic Soy Agar with 10% Sheep Blood) For the cultivation of microorganisms.