Motic Panthera C2 Binocular and Trinocular Microscopes
Supplier: Motic
Certificates
About this item
This new Panthera C2 cased in a revolutionary technical future-orientated solution, now accessible for life sciences.
- Focusable and centerable Abbe condenser N.A. 0.90/1.25 with slot for contrast sliders
- Compact mechanical stage with built-in low position rackless coaxial stage control and sample holder
- Fine focus precision of 2 μm, focusing stroke of 25 mm
- USB 2.0 for external camera power
Ultra-Contrast Optics for best insights, convincing performance of the dual-slide holder for one-hand operation, and intelligent light management with an LED 'Feedback indicator': a setup ready to smoothen daily workflow.
The Panthera C2 is prepared for Phase contrast with slider or turret condenser, Polarization contrast, and Darkfield. It is even possible to integrate a ready-to-go LED Fluorescence module, allowing to apply this advanced method without the multiple problems associated with mercury bulb usage.
The Panthera E2 features optical system, Color Corrected Infinity Optical System, CCIS©.
Delivery information: Dust cover, power cord, Allen key, blue filter, halogen bulb, LED module are included.
Frequently Bought Together