ContactUsLinkComponent
Detecting COVID-19 Using Reverse-Transcription PCR (RT-PCR), Edvotek
Detecting COVID-19 Using Reverse-Transcription PCR (RT-PCR), Edvotek
Catalog # 470336-674
Supplier:  EDVOTEK
undefined
Detecting COVID-19 Using Reverse-Transcription PCR (RT-PCR), Edvotek
Catalog # 470336-674
Supplier:  EDVOTEK
Supplier Number:  363
Specifications
  • Description:
    Detecting COVID-19 using RT-PCR kit
  • No. of lab groups:
    5
  • No. of students:
    25
  • Cat. No.:
    470336-674
Specifications
About this item
In this simulated RT-PCR experiment, students will explore the diagnostic test used worldwide to diagnose and monitor the spread of COVID-19.
  • This kit includes enough materials for 25 students in 5 groups
  • Time required: Set up - 30 minutes; PCR - 2 hours or overnight; Electrophoresis - 45 minutes
Reverse-transcription PCR (RT-PCR) is the gold standard for the detection of SARS-CoV-2 due to the sensitivity, specificity, and feasibility of the test.
Ordering information: Materials required but not included: DNA Electrophoresis, Thermal Cycler, Micropipettes: 5- to 50 µl, UV or Blue Light Transilluminator, White Light Box (optional - use if staining with FlashBlue™), Microcentrifuge and microwave or hot plate.
Materials included: Instructions, PCR EdvoBeads™, LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, DNA Templates, TE Buffer, UltraSpec-Agarose™, TBE Electrophoresis Buffer powder, SYBR® Safe Stain, FlashBlue™ DNA Stain, Microcentrifuge and PCR Tubes.