Pipetting By Numbers: STEAM Pipetting Practice, Edvotek
Catalog # 470336-716
Supplier: EDVOTEK
Specifications
- Description:Pipetting by numbers kit
- No. of lab groups:10
- Cat. No.:470336-716
Specifications
About this item
Help your students master the fundamental biotechnology technique of micro pipetting while creating a colorful, science-themed classroom poster as well as their own artwork.
- Set your classroom up for a year of experimental success by mastering micro pipetting
- Learn best practices, key parts, and the importance of accuracy and precision
- Engage students with a STEAM wet lab based on paint by numbers
- Create a colorful, science-themed classroom poster
This kit includes materials for 10 lab groups and takes approximately 45 minutes to complete.
Ordering information: Materials required but not included: automatic micropipettes with tips, small containers for discarding used tips, scissors, and tape.
Materials included: instructions, dyes, test strips, Pipetting by Numbers canvas and blank canvas, microcentrifuge tubes.
Materials included: instructions, dyes, test strips, Pipetting by Numbers canvas and blank canvas, microcentrifuge tubes.