BioClean-D™ Non Sterile Long Length Hood, Ansell
Catalog # 76521-428
Supplier: Ansell Healthcare
Specifications
- Clothing Size:Universal
- Color:White
- Cat. No.:76521-428
Specifications
About this item
The single-use BioClean-D™ Hood has a three-piece design to ensure a perfect fit.
- Extra-long yoke for maximum coverage
- Lightweight low-linting CleanTough material
- Excellent ESD properties
- Bound seams with single needle stitching
- Polyethylene/non-woven polypropylene laminate construction
Made from antistatic lightweight CleanTough material for comfort, the hood features an extra-long yoke for maximum coverage when worn in conjunction with a coverall with collar and features an elasticated face-opening with reinforced edges to avoid contamination entering the controlled environment.
Certifications: ASTM F739, CE 0598, EN ISO 13688:2013, EN 1149-5:2008, EN 13934-1, EN 19935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325, Category III, EN 13034:2005 + A1:2009.