ContactUsLinkComponent
Fantastik® Multi-Surface Disinfectant Degreaser, Herbal Scent, Essendant
  76500-244
undefined
Fantastik® Multi-Surface Disinfectant Degreaser, Herbal Scent, Essendant
  76500-244
 :  SJN311836
  • Capacity:
    32 oz.
  • Cat. No.:
    76500-244
  • Packaging:
    Spray bottle
Fantastik Multi-Surface Disinfectant Degreaser cleans and fights tough grease and grime.
  • When used as directed on hard, non-porous surfaces, kills 99.9% of viruses and bacteria
  • Removes 100% tough soap scum and grease
  • Leaves a pleasant herbal scent
Kills 99.9% of bacteria, and cuts through tough grease and grime on most surfaces. Clean, disinfect, degrease, and deodorize kitchen and restroom hard, non-porous surfaces. Suitable for use on acrylic, aluminum, bronze, brushed nickel, chrome, fiberglass, quartz, sealed granite, sealed marble, stainless steel, and tile. Chemical compound: Alkyl dimethyl benzyl ammonium chloride (50%C14, 40%C12, 10%C16): 0.26%.