Specifications
- Capacity:32 oz.
- Cat. No.:76500-238
- Packaging:Bottle, pull-top
Specifications
About this item
Healthcare professionals trust Clorox® Bleach Germicidal Cleaner to overcome infection control challenges in a constantly changing healthcare environment. This ready-to-use, one-step cleaner disinfectant is designed for disinfecting surfaces in healthcare settings.
- Ready to use disinfectant that is EPA registered to kill 50 microorganisms
- Powerful cleaner is ideal for use on spills and larger areas
- Use in hospitals, medical facilities, dental offices, veterinary offices, and long term care facilities
- Meets CDC and OSHA recommendations
- An anti-corrosion agent ensures compatibility with healthcare surfaces
- Tested for surface safety on common healthcare and office surfaces and equipment
This cleaner disinfectant is EPA registered to kill microorganisms, including HAI causing bacteria like MRSA and VRE, outbreak causing viruses such as Influenza A and B viruses, Measles Virus, Norovirus, and fungi in one minute. This disinfectant kills 99.99% of C. difficile spores in three minutes. In addition, this cleaner meets EPA criteria for use against SARS-CoV-2, the virus that causes COVID-19. The ready-to-use disinfectant eliminates compliance risks and labor costs associated with dilutable disinfectants. Designed for easy refilling to meet the needs of heavy users.