Spray Nine Heavy-Duty Cleaner/Degreaser/Disinfectant, Citrus Scent, Essendant
Catalog # 76500-228
Supplier: Janitorial Supplies
Specifications
- Capacity:32 oz.
- Cat. No.:76500-228
- Packaging:Spray bottle
Specifications
About this item
A professional-grade cleaner and disinfectant that removes all types of dirt, grease, grime, and stains.
- Professional-grade cleaner, degreaser, and disinfectant
- Disinfects against viruses in 30 seconds
- Disinfect against bacteria in 45 seconds
- Sanitizes in 10 seconds
- Kills fungus in 3 minutes
- Deodorizes
- Controls mold and mildew
- Removes stains
One product with nine distinct uses. This formula cleans, degreases, disinfects against viruses, disinfects against bacteria, sanitizes, kills fungus, deodorizes, controls mold and mildew, and removes stains. Use in bathrooms, breakrooms, kitchens, offices, manufacturing plants, medical offices, and exam rooms. Suitable for use on tables, walls, floors, countertops, applicance exteriors, tubs, showers, glazed tile, toilet exteriors, or garbage pails. Chemical compound: N-Alkyl Dimethyl Benzyl Ammonium Chloride; N-Alkyl Dimethyl Ethybenzyl Ammonium Chloride; Ethoxylated Alcohols; Dipropylene Glycol Mononbutyl Ether.