ContactUsLinkComponent
Diversey™ Glance Glass and Multi-Surface Cleaner, Essendant
Diversey™ Glance Glass and Multi-Surface Cleaner, Essendant
Catalog # 76500-204
undefined
Diversey™ Glance Glass and Multi-Surface Cleaner, Essendant
Catalog # 76500-204
Supplier Number:  DVO04705
Specifications
  • Capacity:
    32 oz.
  • Cat. No.:
    76500-204
  • Packaging:
    Spray bottle
Specifications
About this item
Non-streaking, quick-drying, ammoniated glass cleaner. Cleans and shines glass, mirrors, sinks, countertops, stainless steel, and many other washable surfaces.
  • Concentrated formula delivers excellent cleaning performance
  • Cuts through grease, grime, soap films, finger marks, and smoke film
  • Do not use on copper or brass as repeated use may discolor these surfaces
  • Professional strength formula for jobs big and small
Ready-to-use, spray-and-wipe formula. Suitable for use on ceramic, glass, laminates, metal, plastic, and vinyl. Chemical compound: 2-Butoxyethanol.