ContactUsLinkComponent
Clorox® VersaSure™ Cleaner Disinfectant Wipes, Essendant
Clorox® VersaSure™ Cleaner Disinfectant Wipes, Essendant
Catalog # 76500-374
undefined
Clorox® VersaSure™ Cleaner Disinfectant Wipes, Essendant
Catalog # 76500-374
Supplier Number:  CLO31757
Specifications
  • Color:
    White
  • Cat. No.:
    76500-374
  • Packaging:
    85 wipes/canister
  • W×L:
    6.75×8"
Specifications
About this item
Clorox® Healthcare VersaSure™ Cleaner Disinfectant Wipes kill microorganisms quickly on a broad range of surfaces. These strong and durable alcohol-free wipes are a one-step solution to cleaning and disinfecting.
  • Quaternary ammonium disinfecting wipes
  • Wipes are noncorrosive and contain no added fragrances, chemical odors, fumes, or bleach
  • Disinfect most bacteria and viruses in as little as 30 seconds
VersaSure™ wipes are EPA-registered to kill pathogens on hard surfaces in as fast as 30 seconds for most bacteria and viruses. Designed for patient and staff comfort, these wipes are formulated with no harsh chemical odors, fumes, or added fragrances. Intended for use in commercial settings. Wipes are suitable for use on aluminum, baked enamel, brass, chrome, copper, corian, finished wood, formica, galvanized steel, glass, glazed porcelain, laminate, linoleum, marble, metal, naugahyde, painted metal, painted wood, plastic, plastic laminate, plexiglass, porcelain enamel, rubber, sealed fiberglass, sealed granite, sealed wood, silver, stainless steel, synthetic marble, treated wood, vinyl tile, and wood.