Disinfecting Multi-Surface Wipes, Essendant
76500-310
- Color:White
- Cat. No.:76500-310
- Packaging:200 wipes/container
- W×L:7.5×5.325"
Helps prevent the spread of the cold and flu virus and top foodborne pathogens like Listeria monocytogenes, Campylobacter jejuni, and E. coli. Suitable for use on non-porous surfaces.
- Prevent the spread of the cold and flu virus and top foodborne pathogens
- Use on tables, hand-rails, doorknobs, and other high touch surfaces
Kills 24 different microorganisms commonly found on surfaces in public places.