ContactUsLinkComponent
Oxivir® Five 16 One-Step Disinfectant Cleaner, Essendant
Oxivir® Five 16 One-Step Disinfectant Cleaner, Essendant
Catalog # 76500-298
undefined
Oxivir® Five 16 One-Step Disinfectant Cleaner, Essendant
Catalog # 76500-298
Supplier Number:  DVO4963314
Specifications
  • Capacity:
    1 gal.
  • Cat. No.:
    76500-298
  • Packaging:
    Bottle
Specifications
About this item
Proprietary Accelerated Hydrogen Peroxide (AHP®) based disinfectant-cleaner delivers fast, effective cleaning performance.
  • Hospital-grade disinfectant-cleaner
  • Based on proprietary, Accelerated Hydrogen Peroxide (AHP®) technology
  • Disinfects in 5 minutes at 1:16 dilution ratios
  • Excellent surface safety
  • HMIS ratings of zero
At a 1:16 dilution ratio, product disinfects hard, non-porous surfaces in five minutes. Use as a virucide, bactericide, fungicide, mildewcide, and a non-food contact sanitizer. Kills MRSA, norovirus, and canine parvovirus. Suitable for use on ceramic, fiberglass, glass, laminates, metal, plastic, sealed stone, and vinyl. Chemical compound: 2-Hydroxybenzoic Acid; Phosphoric Acid; Hydrogen Peroxide.