Bond Elut C18, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Bond Elut C18 is a hydrophobic, end capped silica sorbent commonly used to extract nonpolar analytes.
- The most hydrophobic, bonded silica sorbent
- Extremely retentive for nonpolar compounds
- Effective for desalting aqueous mixtures
Bond Elut C18 is the most hydrophobic, bonded silica sorbent available in the Bond Elut family. It is considered to have the broadest retention spectrum among bonded silica sorbents. C18 is the most popular SPE sorbent because of its extreme retention of nonpolar compounds.
Bond Elut C18 retains most organic analytes from aqueous matrices. When analyzing small to intermediate molecules, Bond Elut C18 can be used for desalting aqueous matrices before ion exchange, as salts pass through the sorbent unretained.
Specifications
- Catalog No:
- AG14113001
- AG52102001
- AG52102052
- AG14102052
- AG14256031
- AG12256023
- AG12256001B
- AG14102001
- AG52113024
- AG12256031
- AG14256001
- AG52102024
- AG14256015
- AG14113027
- AG14102028B
- AG12105027
- AG12105102B
- AG14113024
- AG14102028
- AG14102025
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
Specifications
Well plates
Product Information
Frequently Bought Together