Specifications
- Description:EMnetik PCR cleanup kit
- Cat. No.:76518-676
- No. of reactions:500 preparations for 50 µl reactions
Specifications
About this item
DNA cleanup from enzymatic reactions. The EMnetik PCR Cleanup Kit is a revolutionary bead-based DNA cleanup kit. Using new SuperSPRI bead technology combined with the microparticle processor EMnetik 24, a user can complete PCR cleanups in 16 minutes.
- ~2X faster turnaround time compared to column cleanups (16 min vs 30 min)
- >80% recovery (comparable to column cleanup kits)
- Significantly fewer touchpoints (< 50 touchpoints compared to 300 for columns)
- No need to handle small columns or use a single-channel pipette
- Move samples from a thermocycler to the EMnetik 24 system, and don’t move them again until final elution
- Intuitive user interface removes guesswork by providing clear, step-by-step instructions