µ-Slide I Luer Channel Slides, Ibidi
Channel slides with different heights, volumes, and coatings are specifically suited for flow applications.
- Large area of uniform shear stress
- Easy connection using Luer adapters
- Homogeneous cell distribution with defined optical pathway
- Channel dimensions: 5W×50L mm
- Coating area: 5.4 cm²
The µ-Slide I Luer is designed for cell culture under perfusion and all flow applications. Main applications are the simulation of blood vessels for arteriosclerosis research and applying defined shear stress and shear rates on cells inside the channel. The female Luers allow easy connections to tubing and pump systems.
Frequently Bought Together