ContactUsLinkComponent
Clamp for sticky-Slides, Ibidi
Clamp for sticky-Slides, Ibidi
  MSPP-80040
Clamp for sticky-Slides, Ibidi
  MSPP-80040
 :  80040
  • Description:
    Clamp for sticky-Slides
  • Material:
    Aluminum, anodized
  • Cat. No.:
    MSPP-80040
  • Supplier no.:
    80040
This instrument securely fastens sticky-Slides to coverslip bottoms.
  • Applies homogeneous force for tight sealing, with no leakage
  • Clamping force: 1.2 kN
  • Easy to use for an optimal bond; clamp must be paired with one adapter
  • Clamp and adapters must be ordered separately
The clamp creates optimal adhesive bond between sticky-Slide and coverslip to provide a tight seal.
 : Adapters sold seperately.