Coverslips for Sticky-Slides, Ibidi
Supplier: IBIDI USA MS
Certificates
About this item
Coverslips for ibidi sticky-slides, available in glass or polymer formats.
- Polymer coverslip available with ibiTreat tissue culture treatment, or uncoated
- Glass coverslip made from high-quality D 263 M Schott glass
Coverslips for sticky-Slides, designed for high–end microscopic analysis with uncompromised resolution and choice of wavelength.
Frequently Bought Together