Au-Flat Grids, Electron Microscopy Sciences
About this item
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
- Holey Au/Pd film on a gold mesh grid
- Stronger than carbon films
- More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
3 Options Available Below
Product Details & Documents
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
- Holey Au/Pd film on a gold mesh grid
- Stronger than carbon films
- More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
Product Family Options
Product Information
- MaterialHole sizeAvailabilityPrice
- 76499-686Au-flat cryo-em grid, 200 µm mesh sizeAu-flat cryo-em grid, 200 µm mesh sizeGold2 µm (2 µm spacing)Log in for availabilityPack of 50$1,683.40Supplier Number: AUFT222-50
- 76499-688Au-flat cryo-em grid, 300 µm mesh sizeAu-flat cryo-em grid, 300 µm mesh sizeGold1.2 µm (1.3 µm spacing)Log in for availabilityPack of 5$228.36Supplier Number: AUFT313-05
- 76499-690Au-flat cryo-em grid, 300 µm mesh sizeAu-flat cryo-em grid, 300 µm mesh sizeGold1.2 µm (1.3 µm spacing)Log in for availabilityPack of 50$1,683.40Supplier Number: AUFT313-50