Brady® High Adhesion Clear Polyester Product Identification Labels, Brady
Supplier: Brady Worldwide
Certificates
About this item
Rough surface polyester (B-432) labels are ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
- Recommended ribbons: B30-R4400-WT;B30-R6000
- Heat resistant label with permanent adhesive
- Permanent acrylic adhesive type
- Polyester material
Made from durable, glossy white polyester with permanent acrylic adhesive. Chemical, oil, and water resistant. UL Recognized for outdoor use on glass, thermoset polyester plastic, and polyvinyl fluoride plastic surfaces to support solar panel identification. Service temperature of −70 to 230 °F (−56 to 110 °C). A ribbon is required to print this material.
Frequently Bought Together