Brady®, B33 Series PermaShield™ Glossy White Polyester Labels, Brady
Supplier: Brady Worldwide
Certificates
About this item
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, barcode labels, rating plates, and solar panel identification.
- Recommended ribbon: B30-R6000
- Harsh environment, heat resistant label with permanent adhesive
- Permanent acrylic adhesive type
- Polyester with polyester overlaminate material
- UL approved, CSA accepted
Brady PermaShield™ labels are supplied as a two-in-one label comprised of a standard printable label with matching clear polyester overlaminate mounted beneath the label. These labels meet the requirements of a halogen-free material per DIN VDE 0472 part 815. Service temperature of −94 to 230 °F (−70 to 110 °C). A ribbon is required to print this material.
Frequently Bought Together