About this item
B-2475 polyester material meets UL 969 standard for marking and labeling, and is ideal for product labeling, logos, Right to Know labels, and barcode applications.
- Recommended ribbon: J20-CMY
- Chemical resistant labels with permanent adhesive
- Acrylic adhesive type
- Inkjet receptive polyester material
Add color and edit labels on demand for a no-waste, low-inventory labeling solution. Full-color printing allows users to create labels with variable data and color components in house. Durable and resistant to abrasions, chemicals, oils, and solvents. This inkjet-receptive polyester with acrylic adhesive works best on flat, smooth surfaces. Reduces label stocking as labels are created and printed on demand. Decreases waste as labels can be reviewed and revised immediately if errors are found, unlike pre-printed labels, which require label data to be set in advance. Allows for immediate in-house label printing, eliminates stress associated with long lead times, and reduces the possibility of experiencing a line down due to label shortages. An ink cartridge is required to print this material. The recommended ribbon is J20-CMY.